siRNA Details
Gene Symbol: GPR162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014449.1 | NP_055264.1 | 5 | 1178 | ||
NM_019858.1 | NP_062832.1 | 5 | 2180 | |||
GenBank mRNA | AB209836.1 | - | 4 | 1587 | ||
BC002353.2 | - | 5 | 1272 | 2821031 | ||
BC007918.2 | - | 4 | 811 | |||
BC080585.1 | - | 5 | 1159 | 3510624 | ||
U47925.1 | - | 5 | 1178 | |||
U47928.1 | - | 5 | 2180 |
