siRNA Details
| Gene Symbol: SULT1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014465.3 | NP_055280.2 | 3, 4 | 575 | ||
| XM_005265676.3 | XP_005265733.1 | 4, 5 | 490 | |||
| XM_005265677.3 | XP_005265734.1 | 4, 5 | 690 | |||
| XM_005265678.3 | XP_005265735.1 | 3, 4 | 443 | |||
| XM_011531873.2 | XP_011530175.1 | 2, 3 | 314 | |||
| GenBank mRNA | BC010895.1 | - | 3, 4 | 577 | 4105139 | |
| D89479.1 | - | 2, 3 | 301 | |||
| U95726.1 | - | 2, 3 | 292 | |||

SULT1B1
Genomic Map