siRNA Details
| Gene Symbol: CD22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185099.1 | NP_001172028.1 | 4 | 769 | ||
| NM_001185100.1 | NP_001172029.1 | 4 | 769 | |||
| NM_001185101.1 | NP_001172030.1 | 4 | 769 | |||
| NM_001278417.1 | NP_001265346.1 | 3 | 597 | |||
| NM_001771.3 | NP_001762.2 | 4 | 769 | |||
| GenBank mRNA | AK026467.1 | - | 2 | 1594 | ||
| AK225625.1 | - | 4 | 751 | |||
| AK225694.1 | - | 4 | 751 | |||
| AK301177.1 | - | 4 | 632 | |||
| BC109306.1 | - | 3 | 551 | 40035425 | ||
| X52785.1 | - | 3 | 719 | |||
| X59350.1 | - | 4 | 741 | |||

CD22
Genomic Map