siRNA Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292002.1 | NP_001278931.1 | 2 | 280 | ||
| NM_014487.5 | NP_055302.1 | 2 | 280 | |||
| XM_017008033.1 | XP_016863522.1 | 2 | 424 | |||
| GenBank mRNA | AJ006591.2 | - | 2 | 225 | ||
| AK002106.1 | - | 2 | 230 | |||
| AK223609.1 | - | 2 | 231 | |||
| AK294089.1 | - | 2 | 234 | |||
| AK315463.1 | - | 2 | 266 | |||
| BC004421.1 | - | 2 | 256 | 3352916 | ||
| BU532661.1 | - | 2 | 283 | |||
| DA089047.1 | - | 2 | 272 | |||

ZNF330
Genomic Map