siRNA Details
| Gene Symbol: BHLHE22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152414.4 | NP_689627.1 | 1 | 2512 | ||
| GenBank mRNA | AK091333.1 | - | 1 | 1586 | ||
| BC033056.1 | - | 1 | 365 | |||
| BC045748.1 | - | 1 | 367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BHLHE22| Gene Symbol: BHLHE22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152414.4 | NP_689627.1 | 1 | 2512 | ||
| GenBank mRNA | AK091333.1 | - | 1 | 1586 | ||
| BC033056.1 | - | 1 | 365 | |||
| BC045748.1 | - | 1 | 367 | |||