siRNA Details
| Gene Symbol: EIF3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308393.1 | NP_001295322.1 | 5, 6 | 601 | ||
| NM_013234.3 | NP_037366.1 | 5, 6 | 609 | |||
| GenBank mRNA | AB019392.1 | - | 5, 6 | 585 | ||
| AF077051.1 | - | 5, 6 | 585 | |||
| AF085358.1 | - | 5, 6 | 586 | |||
| AF109355.1 | - | 5, 6 | 475 | |||
| AF315506.1 | - | 5, 6 | 417 | |||
| AK026531.1 | - | 5, 6 | 456 | |||
| AK289554.1 | - | 5, 6 | 487 | |||
| AK298635.1 | - | 4, 5 | 388 | |||
| AK301035.1 | - | 5, 6 | 489 | |||
| AK316344.1 | - | 5, 6 | 494 | |||
| AY245432.1 | - | 5, 6 | 579 | |||
| BC001031.2 | - | 5, 6 | 473 | 3139375 | ||
| BC007335.2 | - | 2, 3 | 111 | |||
| BC007559.1 | - | 5, 6 | 500 | 2988566 | ||
| BF965565.1 | - | 5, 6 | 437 | |||
| BG488547.1 | - | 5, 6 | 472 | |||
| JX870647.1 | - | 4, 5 | 502 | |||

EIF3K
Genomic Map