siRNA Details
| Gene Symbol: UBE2S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014501.2 | NP_055316.2 | 2 | 471 | ||
| XM_011526752.1 | XP_011525054.1 | 2 | 660 | |||
| GenBank mRNA | AB062397.1 | - | 1 | 281 | ||
| BC004236.2 | - | 1 | 281 | 2988449 | ||
| BC007554.1 | - | 1 | 281 | 2967547 | ||
| BC065364.1 | - | 1 | 291 | 5434942 | ||
| BC066948.1 | - | 1 | 127 | 4836851 | ||
| M91670.1 | - | 2 | 162 | |||

UBE2S
Genomic Map