siRNA Details
| Gene Symbol: TOR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317893.1 | NP_001304822.1 | 3 | 632 | ||
| NM_014506.2 | NP_055321.1 | 3 | 632 | |||
| NR_133948.1 | 3 | 773 | ||||
| NR_134113.1 | 3 | 773 | ||||
| XM_005251927.3 | XP_005251984.1 | 3 | 627 | |||
| GenBank mRNA | AF007872.1 | - | 2 | 353 | ||
| AF317129.1 | - | 3 | 622 | |||
| AI568476.1 | - | 2 | 329 | |||
| BC015578.2 | - | 3 | 577 | 4589479 | ||

TOR1B
Genomic Map