siRNA Details
Gene Symbol: MAT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013283.4 | NP_037415.1 | 3 | 486 | ||
NM_182796.2 | NP_877725.1 | 3 | 625 | |||
GenBank mRNA | AB073390.1 | - | 3 | 400 | ||
AF113225.1 | - | 3 | 282 | |||
AF182814.1 | - | 3 | 327 | |||
AJ243721.1 | - | 3 | 465 | |||
AK096178.1 | - | 2 | 1746 | |||
AK291919.1 | - | 3 | 436 | |||
AK312365.1 | - | 3 | 435 | |||
AL136664.1 | - | 3 | 399 | |||
AU279489.1 | - | 3 | 435 | |||
AY358695.1 | - | 3 | 380 | |||
BC005218.1 | - | 3 | 406 | 3931953 | ||
BC066645.1 | - | 3 | 361 | 5749043 | ||
BC093030.1 | - | 3 | 395 | 30527606 | ||
DA146198.1 | - | 3 | 489 | |||
DQ413183.1 | - | 3 | 464 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
