siRNA Details
| Gene Symbol: CECR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290046.1 | NP_001276975.1 | 16 | 2246 | ||
| NM_001290047.1 | NP_001276976.1 | 16 | 2715 | |||
| XM_006724079.3 | XP_006724142.1 | 4 | 1158 | |||
| XM_011546128.2 | XP_011544430.1 | 15 | 2319 | |||
| XM_011546129.2 | XP_011544431.1 | 16 | 2319 | |||
| XM_011546132.2 | XP_011544434.1 | 16 | 2403 | |||
| XM_017028785.1 | XP_016884274.1 | 16 | 2403 | |||
| GenBank mRNA | AB051527.1 | - | 8 | 1264 | ||
| AF336133.1 | - | 16 | 2775 | |||
| AK308330.1 | - | 16 | 2247 | |||
| AL832377.1 | - | 16 | 2193 | |||
| BX647449.1 | - | 16 | 2193 | |||

CECR2
Genomic Map