siRNA Details
| Gene Symbol: SLCO4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016354.3 | NP_057438.3 | 10 | 2048 | ||
| XM_005260203.2 | XP_005260260.1 | 10 | 2045 | |||
| XM_011528792.2 | XP_011527094.1 | 10 | 2045 | |||
| XM_017027826.1 | XP_016883315.1 | 10 | 2029 | |||
| XM_017027827.1 | XP_016883316.1 | 10 | 2045 | |||
| GenBank mRNA | AB031051.1 | - | 9 | 1934 | ||
| AF104334.1 | - | 8 | 2372 | |||
| AF187817.1 | - | 9 | 1843 | |||
| AF205072.1 | - | 10 | 1988 | |||
| AK000551.1 | - | 2 | 130 | |||
| AK023410.1 | - | 6 | 1549 | |||
| BC015727.1 | - | 10 | 2006 | 4908245 | ||

SLCO4A1
Genomic Map