siRNA Details
| Gene Symbol: TRIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021643.3 | NP_067675.1 | 1 | 1600 | ||
| GenBank mRNA | AK313237.1 | - | 1 | 650 | ||
| AY245544.1 | - | 1 | 1583 | |||
| BC002637.2 | - | 1 | 650 | 3607549 | ||
| D87119.1 | - | 1 | 1389 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_144551, AK082329, BC037387, AK144192, AF358867, and others.

TRIB2
Genomic Map