siRNA Details
| Gene Symbol: DEXI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014015.3 | NP_054734.2 | 2 | 1037 | ||
| XM_011522455.2 | XP_011520757.1 | 3 | 419 | |||
| XM_017023171.1 | XP_016878660.1 | 3 | 1805 | |||
| GenBank mRNA | AF108145.2 | - | 2 | 653 | ||
| BC037569.1 | - | 2 | 1037 | 5227606 | ||
| BC052989.1 | - | 2 | 770 | 6194689 | ||
| BC071683.1 | - | 2 | 936 | 5190179 | ||

DEXI
Genomic Map