siRNA Details
| Gene Symbol: SLC27A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017372.2 | NP_001017372.1 | 8, 9 | 2561 | ||
| NM_001317984.1 | NP_001304913.1 | 9, 10 | 2385 | |||
| NM_014031.4 | NP_054750.1 | 9, 10 | 2336 | |||
| GenBank mRNA | AF064254.1 | - | 8, 9 | 1703 | ||
| AK313456.1 | - | 9, 10 | 1984 | |||
| AL833117.1 | - | 8, 9 | 2134 | |||
| BC041945.1 | - | 8, 9 | 2560 | 5301306 | ||
| BX537383.1 | - | 9, 10 | 1946 | |||
| CR457129.1 | - | 8, 9 | 1550 | |||

SLC27A6
Genomic Map