siRNA Details
| Gene Symbol: SNX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014035.2 | NP_054754.1 | 3 | 397 | ||
| XM_005271972.4 | XP_005272029.1 | 3 | 286 | |||
| XM_006714592.3 | XP_006714655.1 | 3 | 352 | |||
| XM_011543349.2 | XP_011541651.1 | 4 | 417 | |||
| XM_011543350.2 | XP_011541652.1 | 4 | 416 | |||
| XM_011543352.1 | XP_011541654.1 | 4 | 417 | |||
| XM_017009394.1 | XP_016864883.1 | 3 | 222 | |||
| XM_017009395.1 | XP_016864884.1 | 4 | 417 | |||
| XM_017009396.1 | XP_016864885.1 | 4 | 417 | |||
| GenBank mRNA | AF139461.1 | - | 3 | 286 | ||
| AY044655.1 | - | 3 | 228 | |||
| AY358098.1 | - | 3 | 246 | |||
| BC010886.1 | - | 3 | 357 | 3538426 | ||
| BC069012.1 | - | 3 | 344 | 4585781 | ||
| BQ278046.1 | - | 3 | 369 | |||

SNX24
Genomic Map