siRNA Details
| Gene Symbol: AAMDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316957.1 | NP_001303886.1 | 3 | 248 | ||
| NM_001316958.1 | NP_001303887.1 | 3 | 248 | |||
| NM_001316960.1 | NP_001303889.1 | 3 | 324 | |||
| NM_001316961.1 | NP_001303890.1 | 3 | 244 | |||
| NM_001316962.1 | NP_001303891.1 | 3 | 248 | |||
| NM_024684.3 | NP_078960.1 | 2 | 206 | |||
| NR_133637.1 | 2 | 206 | ||||
| XM_011544968.2 | XP_011543270.1 | 2 | 206 | |||
| XM_011544969.2 | XP_011543271.1 | 2 | 199 | |||
| XM_017017620.1 | XP_016873109.1 | 2 | 228 | |||
| GenBank mRNA | AF092136.1 | - | 3 | 207 | ||
| AK024688.1 | - | 2 | 165 | |||
| BC002752.2 | - | 2 | 158 | 3628061 | ||
| BC016854.2 | - | 2 | 135 | 3856546 | ||
| BI791786.1 | - | 3 | 257 | |||
| BM930331.1 | - | 3 | 193 | |||
| BP270361.1 | - | 2 | 206 | |||
| BX646013.1 | - | 2 | 190 | |||
| CN362204.1 | - | 3 | 219 | |||
| CR457332.1 | - | 1 | 60 | |||

AAMDC
Genomic Map