siRNA Details
| Gene Symbol: SPCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014041.3 | NP_054760.3 | 3 | 554 | ||
| GenBank mRNA | AF092138.1 | - | 3 | 283 | ||
| AK027426.1 | - | 3 | 283 | |||
| AX775783.1 | - | 3 | 283 | |||
| AX775785.1 | - | 4 | 1068 | |||
| AX775787.1 | - | 3 | 257 | |||
| AX775833.1 | - | 3 | 273 | |||
| BC000884.1 | - | 3 | 257 | 3462307 | ||
| BI826431.1 | - | 3 | 248 | |||
| BQ423345.1 | - | 3 | 290 | |||
| L38852.1 | - | 3 | 273 | |||

SPCS1
Genomic Map