siRNA Details
| Gene Symbol: TNFSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244.3 | NP_001235.1 | 4 | 633 | ||
| NM_001252290.1 | NP_001239219.1 | 4 | 633 | |||
| GenBank mRNA | AK313619.1 | - | 4 | 633 | ||
| AL583443.3 | - | 4 | 537 | |||
| BC093630.1 | - | 4 | 415 | 7939475 | ||
| BC111939.1 | - | 4 | 415 | 8327407 | ||
| CD690231.1 | - | 4 | 343 | |||
| L09753.1 | - | 4 | 455 | |||

TNFSF8
Genomic Map