siRNA Details
| Gene Symbol: MRPS18B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014046.3 | NP_054765.1 | 2 | 282 | ||
| GenBank mRNA | AF100761.1 | - | 2 | 137 | ||
| AF151017.1 | - | 2 | 163 | |||
| AK294086.1 | - | 2 | 144 | |||
| AK311890.1 | - | 2 | 133 | |||
| AL050361.1 | - | 2 | 141 | |||
| BC005373.1 | - | 2 | 162 | 3933607 | ||
| BG773408.1 | - | 2 | 273 | |||
| BP369345.1 | - | 2 | 282 | |||

MRPS18B
Genomic Map