siRNA Details
| Gene Symbol: C19orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014047.2 | NP_054766.1 | 3 | 216 | ||
| GenBank mRNA | AF078852.1 | - | 3 | 190 | ||
| AF086483.1 | - | 3 | 188 | |||
| AK311855.1 | - | 3 | 181 | |||
| BC015465.2 | - | 3 | 191 | 3914049 | ||
| DA768829.1 | - | 3 | 217 | |||

C19orf53
Genomic Map