siRNA Details
| Gene Symbol: TMEM14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014051.3 | NP_054770.1 | 2 | 228 | ||
| GenBank mRNA | AF078864.1 | - | 2 | 272 | ||
| AF239771.1 | - | 2 | 207 | |||
| AK312063.1 | - | 2 | 201 | |||
| BC015097.1 | - | 2 | 187 | 3686196 | ||
| BC019328.1 | - | 2 | 183 | 3534851 | ||
| CB117714.1 | - | 2 | 228 | |||

TMEM14A
Genomic Map