siRNA Details
| Gene Symbol: TMEM14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014051.3 | NP_054770.1 | 3, 4 | 341 | ||
| GenBank mRNA | AF078864.1 | - | 3, 4 | 385 | ||
| AF239771.1 | - | 3, 4 | 320 | |||
| AK312063.1 | - | 3, 4 | 314 | |||
| BC015097.1 | - | 3, 4 | 300 | 3686196 | ||
| BC019328.1 | - | 3, 4 | 296 | 3534851 | ||

TMEM14A
Genomic Map