siRNA Details
| Gene Symbol: ATAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014109.3 | NP_054828.2 | 8 | 1101 | ||
| XM_011516994.2 | XP_011515296.1 | 8 | 1628 | |||
| XM_011516995.2 | XP_011515297.1 | 8 | 1628 | |||
| GenBank mRNA | AK097133.1 | - | 8 | 1083 | ||
| AL833653.1 | - | 8 | 855 | |||
| AY598335.1 | - | 8 | 993 | |||
| BC113656.1 | - | 8 | 1015 | 8322708 | ||
| CR749832.1 | - | 9 | 1357 | |||

ATAD2
Genomic Map