siRNA Details
| Gene Symbol: ZC3H7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014153.3 | NP_054872.2 | 17, 18 | 2252 | ||
| XM_006720877.2 | XP_006720940.1 | 17, 18 | 2288 | |||
| XM_011522463.1 | XP_011520765.1 | 18, 19 | 2437 | |||
| XM_011522464.2 | XP_011520766.1 | 11, 12 | 2045 | |||
| XM_017023174.1 | XP_016878663.1 | 17, 18 | 2269 | |||
| XM_017023175.1 | XP_016878664.1 | 13, 14 | 1882 | |||
| GenBank mRNA | AK000325.1 | - | 15, 16 | 1991 | ||
| AK026827.1 | - | 4, 5 | 437 | |||
| AK026956.1 | - | 9, 10 | 1477 | |||
| AK091803.1 | - | 15, 16 | 1954 | |||
| AY163807.1 | - | 16, 17 | 2281 | |||
| BC046363.1 | - | 17, 18 | 2145 | 5538820 | ||

ZC3H7A
Genomic Map