siRNA Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014167.4 | NP_054886.2 | 2 | 212 | ||
| NR_033192.1 | 2 | 141 | ||||
| GenBank mRNA | AF161477.1 | - | 2 | 186 | ||
| AF213377.1 | - | 2 | 179 | |||
| AK001156.1 | - | 2 | 188 | |||
| AK225139.1 | - | 2 | 191 | |||
| BC020647.1 | - | 2 | 175 | 4771954 | ||
| BM846580.1 | - | 2 | 111 | |||
| DA666239.1 | - | 2 | 141 | |||

CCDC59
Genomic Map