siRNA Details
| Gene Symbol: METTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293186.1 | NP_001280115.1 | 3 | 399 | ||
| NM_001293187.1 | NP_001280116.1 | 3 | 313 | |||
| NM_014168.3 | NP_054887.2 | 2 | 540 | |||
| XM_011511023.1 | XP_011509325.1 | 3 | 340 | |||
| GenBank mRNA | AF161482.1 | - | 3 | 288 | ||
| AF201938.1 | - | 3 | 357 | |||
| AK001321.1 | - | 3 | 287 | |||
| BC000921.2 | - | 2 | 452 | 3445582 | ||
| BC093014.1 | - | 3 | 295 | 4799279 | ||
| CA337169.1 | - | 3 | 314 | |||

METTL5
Genomic Map