siRNA Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014169.3 | NP_054888.2 | 3, 4 | 756 | ||
| GenBank mRNA | AB100262.2 | - | 3, 4 | 391 | ||
| AF161483.1 | - | 3, 4 | 410 | |||
| AF212243.1 | - | 3, 4 | 386 | |||
| AY329084.1 | - | 3, 4 | 365 | |||
| BC010893.1 | - | 3, 4 | 393 | |||
| BC107699.1 | - | 3, 4 | 418 | |||
| BC113533.1 | - | 3, 4 | 756 | 8322585 | ||
| BC113535.1 | - | 3, 4 | 756 | 8322587 | ||

CHMP4A
Genomic Map