siRNA Details
| Gene Symbol: MRPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014175.3 | NP_054894.1 | 5 | 903 | ||
| GenBank mRNA | AF161494.1 | - | 5 | 895 | ||
| AK312832.1 | - | 5 | 867 | |||
| AL136665.1 | - | 5 | 873 | |||
| BC000891.1 | - | 5 | 903 | 3463430 | ||
| CR533531.1 | - | 5 | 840 | |||
| DA504537.1 | - | 1 | 12 | |||

MRPL15
Genomic Map