siRNA Details
| Gene Symbol: MRPL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014990.2 | NP_001014990.1 | 6 | 398 | ||
| NM_014180.3 | NP_054899.2 | 7 | 516 | |||
| GenBank mRNA | AF161507.1 | - | 7 | 484 | ||
| AK000601.1 | - | 7 | 503 | |||
| AK097880.1 | - | 3 | 2153 | |||
| AK302784.1 | - | 6 | 378 | |||
| BC012565.1 | - | 7 | 486 | 4333117 | ||
| BP372420.1 | - | 7 | 516 | |||
| BU943770.1 | - | 7 | 509 | |||
| CR933679.1 | - | 6 | 377 | |||

MRPL22
Genomic Map