siRNA Details
| Gene Symbol: MRPL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014990.2 | NP_001014990.1 | 5 | 318 | ||
| NM_014180.3 | NP_054899.2 | 6 | 436 | |||
| GenBank mRNA | AF161507.1 | - | 6 | 404 | ||
| AK000601.1 | - | 6 | 423 | |||
| AK097880.1 | - | 2 | 2073 | |||
| AK302784.1 | - | 5 | 298 | |||
| BC012565.1 | - | 6 | 406 | 4333117 | ||
| BP372420.1 | - | 6 | 436 | |||
| BU943770.1 | - | 6 | 429 | |||
| CR933679.1 | - | 5 | 297 | |||

MRPL22
Genomic Map