siRNA Details
| Gene Symbol: CNIH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277199.1 | NP_001264128.1 | 3, 4 | 636 | ||
| NM_001277200.1 | NP_001264129.1 | 3, 4 | 358 | |||
| NM_014184.3 | NP_054903.1 | 3, 4 | 358 | |||
| NR_102347.1 | 2, 3 | 339 | ||||
| GenBank mRNA | AF161512.1 | - | 3, 4 | 277 | ||
| AK024569.1 | - | 273 | ||||
| AK222634.1 | - | 3, 4 | 263 | |||
| AK312064.1 | - | 3, 4 | 283 | |||
| BC000573.1 | - | 3, 4 | 273 | 3163724 | ||
| BQ051754.1 | - | 3, 4 | 547 | |||
| DA185135.1 | - | 3, 4 | 359 | |||
| HY018269.1 | - | 2, 3 | 255 | |||
| HY145928.1 | - | 3, 4 | 283 | |||

CNIH4
Genomic Map