siRNA Details
| Gene Symbol: TMEM208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318217.1 | NP_001305146.1 | 5 | 589 | ||
| NM_014187.3 | NP_054906.2 | 5 | 472 | |||
| NR_134524.1 | 5 | 498 | ||||
| GenBank mRNA | AF161519.1 | - | 5 | 494 | ||
| BC003080.2 | - | 5 | 445 | 3050277 | ||
| BC013412.2 | - | 5 | 424 | 3030307 | ||
| BC021109.1 | - | 5 | 454 | 3660106 | ||
| BF316569.1 | - | 5 | 398 | |||
| BG825054.1 | - | 5 | 550 | |||
| BI833541.1 | - | 5 | 457 | |||

TMEM208
Genomic Map