siRNA Details
| Gene Symbol: DNAJC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013238.2 | NP_037370.2 | 3, 4 | 631 | ||
| GenBank mRNA | AF126743.1 | - | 4, 5 | 653 | ||
| AK311866.1 | - | 3, 4 | 267 | |||
| AY248898.1 | - | 3, 4 | 631 | |||
| AY512565.1 | - | 3, 4 | 324 | |||
| BC010910.1 | - | 3, 4 | 360 | 4332619 | ||
| BC095400.1 | - | 3, 4 | 250 | 30530999 | ||

DNAJC15
Genomic Map