siRNA Details
| Gene Symbol: CTNNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127384.2 | NP_001120856.1 | 9 | 1291 | ||
| NM_013266.3 | NP_037398.2 | 9 | 1338 | |||
| XM_017016151.1 | XP_016871640.1 | 10 | 1333 | |||
| XM_017016152.1 | XP_016871641.1 | 12 | 1630 | |||
| XM_017016153.1 | XP_016871642.1 | 10 | 1341 | |||
| XM_017016154.1 | XP_016871643.1 | 5 | 709 | |||
| XM_017016155.1 | XP_016871644.1 | 5 | 719 | |||
| XM_017016156.1 | XP_016871645.1 | 5 | 596 | |||
| XM_017016157.1 | XP_016871646.1 | 3 | 746 | |||
| GenBank mRNA | AF091606.1 | - | 9 | 1316 | ||
| AK289756.1 | - | 9 | 1227 | |||
| AK313882.1 | - | 9 | 1341 | |||
| BC065819.1 | - | 9 | 1234 | 6189390 | ||

CTNNA3
Genomic Map