siRNA Details
| Gene Symbol: PRSS50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013270.4 | NP_037402.1 | 5, 6 | 977 | ||
| GenBank mRNA | AF100707.1 | - | 5, 6 | 977 | ||
| BC033016.1 | - | 5, 6 | 968 | 4822455 | ||
| BC037775.2 | - | 5, 6 | 939 | 5272458 | ||

PRSS50
Genomic Map