siRNA Details
| Gene Symbol: CD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040031.1 | NP_001035120.1 | 7, 8 | 858 | ||
| NM_001774.2 | NP_001765.1 | 7, 8 | 892 | |||
| XM_011527544.2 | XP_011525846.1 | 7, 8 | 929 | |||
| XM_017027513.1 | XP_016883002.1 | 7, 8 | 890 | |||
| GenBank mRNA | AK058093.1 | - | 8, 9 | 1239 | ||
| BC106752.1 | - | 7, 8 | 809 | 40023348 | ||
| BC110379.1 | - | 7, 8 | 844 | 5752290 | ||
| BT019851.1 | - | 7, 8 | 771 | |||
| CR541877.1 | - | 7, 8 | 771 | |||
| X14046.1 | - | 7, 8 | 834 | |||

CD37
Genomic Map