siRNA Details
| Gene Symbol: CARD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014550.3 | NP_055365.2 | 8 | 1408 | ||
| GenBank mRNA | AK074906.1 | - | 5 | 638 | ||
| AY028896.1 | - | 8 | 1445 | |||
| AY032928.1 | - | 8 | 1408 | |||
| BC032058.1 | - | 6 | 1033 | |||
| BC113659.1 | - | 8 | 1426 | 8322711 | ||
| BC117403.1 | - | 8 | 1420 | 40125954 | ||

CARD10
Genomic Map