siRNA Details
| Gene Symbol: ABT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013375.3 | NP_037507.1 | 1, 2 | 290 | ||
| GenBank mRNA | AB027258.1 | - | 1, 2 | 270 | ||
| BC048812.1 | - | 1, 2 | 288 | 5574755 | ||
| BC066313.1 | - | 1, 2 | 280 | 5272771 | ||

ABT1
Genomic Map