siRNA Details
| Gene Symbol: NCAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185011.1 | NP_001171940.1 | 2, 3 | 340 | ||
| NM_014551.4 | NP_055366.3 | 2, 3 | 340 | |||
| NM_152299.3 | NP_689512.2 | 2, 3 | 340 | |||
| XM_005261912.4 | XP_005261969.1 | 3, 4 | 392 | |||
| XM_011530685.2 | XP_011528987.1 | 2, 3 | 220 | |||
| XM_017028793.1 | XP_016884282.1 | 3, 4 | 378 | |||
| GenBank mRNA | BC000473.2 | - | 2, 3 | 313 | ||
| BC001509.2 | - | 2, 3 | 313 | |||
| BC001833.2 | - | 2, 3 | 313 | |||
| BC001937.1 | - | 2, 3 | 297 | 2819717 | ||
| BC009441.1 | - | 2, 3 | 296 | 3509636 | ||
| CR456604.1 | - | 2, 3 | 272 | |||
| DB498781.1 | - | 2, 3 | 341 | |||

NCAPH2
Genomic Map