siRNA Details
| Gene Symbol: YPEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013313.4 | NP_037445.1 | 5 | 686 | ||
| NR_130910.1 | 5 | 1070 | ||||
| GenBank mRNA | AB098734.1 | - | 5 | 686 | ||
| AB098735.1 | - | 5 | 686 | |||
| AF060862.1 | - | 4 | 386 | |||
| AF305069.1 | - | 4 | 446 | |||
| BC034486.1 | - | 5 | 544 | 4826997 | ||
| BC074501.1 | - | 5 | 551 | 5171826 | ||
| CR456376.2 | - | 4 | 333 | |||
| CR933630.1 | - | 4 | 4789 | |||

YPEL1
Genomic Map