siRNA Details
Gene Symbol: GPR171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013308.3 | NP_037440.3 | 3 | 937 | ||
XM_005247402.3 | XP_005247459.1 | 2 | 1321 | |||
XM_005247403.3 | XP_005247460.1 | 2 | 972 | |||
XM_017006274.1 | XP_016861763.1 | 3 | 1414 | |||
GenBank mRNA | AF002986.1 | - | 3 | 916 | ||
BC036815.1 | - | 3 | 928 | 5747378 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
