siRNA Details
| Gene Symbol: NME7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013330.4 | NP_037462.1 | 10 | 1187 | ||
| NM_197972.2 | NP_932076.1 | 10 | 1156 | |||
| NR_104229.1 | 10 | 1187 | ||||
| XM_005245106.4 | XP_005245163.1 | 8 | 765 | |||
| XM_017001099.1 | XP_016856588.1 | 10 | 1173 | |||
| XM_017001100.1 | XP_016856589.1 | 8 | 753 | |||
| GenBank mRNA | AF153191.2 | - | 10 | 1026 | ||
| AK094513.1 | - | 10 | 1139 | |||
| AK290701.1 | - | 10 | 1021 | |||
| AK301917.1 | - | 10 | 1170 | |||
| BC006983.1 | - | 10 | 1018 | 3997152 | ||

NME7
Genomic Map