siRNA Details
| Gene Symbol: ALG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013339.3 | NP_037471.2 | 12, 13 | 1370 | ||
| GenBank mRNA | AF102851.1 | - | 12, 13 | 1303 | ||
| AK022700.1 | - | 12, 13 | 1308 | |||
| BC001253.1 | - | 12, 13 | 1302 | 3452687 | ||
| CB215801.1 | - | 4, 5 | 296 | |||

ALG6
Genomic Map