siRNA Details
| Gene Symbol: PKN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317926.1 | NP_001304855.1 | 10 | 1707 | ||
| NM_013355.4 | NP_037487.2 | 10 | 1707 | |||
| XM_005251946.3 | XP_005252003.1 | 10 | 2262 | |||
| XM_006717080.2 | XP_006717143.1 | 10 | 1776 | |||
| XM_017014649.1 | XP_016870138.1 | 10 | 1778 | |||
| XM_017014650.1 | XP_016870139.1 | 10 | 1916 | |||
| GenBank mRNA | AB019692.1 | - | 10 | 1295 | ||
| BC062558.1 | - | 10 | 1310 | 6649806 | ||

PKN3
Genomic Map