siRNA Details
| Gene Symbol: SERTAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013368.3 | NP_037500.2 | 2 | 753 | ||
| NM_203344.2 | NP_976219.1 | 2 | 642 | |||
| XM_005258832.3 | XP_005258889.1 | 3 | 850 | |||
| XM_006723179.3 | XP_006723242.1 | 2 | 702 | |||
| GenBank mRNA | AF192529.1 | - | 2 | 766 | ||
| AK074604.1 | - | 2 | 567 | |||
| BC014061.2 | - | 2 | 566 | 3926937 | ||
| BC050643.2 | - | 2 | 642 | 6503515 | ||

SERTAD3
Genomic Map