siRNA Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182981.2 | NP_892026.1 | 6 | 1775 | ||
| GenBank mRNA | AF191740.1 | - | 4 | 1409 | ||
| AY037158.1 | - | 6 | 1740 | |||
| AY258066.1 | - | 7 | 2226 | |||
| AY258067.1 | - | 6 | 2066 | |||
| AY258068.1 | - | 6 | 1756 | |||
| BC020434.1 | - | 6 | 1741 | |||
| BC032476.1 | - | 6 | 1740 | 5215262 | ||

OSGIN1
Genomic Map