siRNA Details
| Gene Symbol: CDIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199054.1 | NP_001185983.1 | 6 | 1546 | ||
| NM_001199055.1 | NP_001185984.1 | 6 | 1397 | |||
| NM_001199056.1 | NP_001185985.1 | 6 | 1277 | |||
| NM_013399.2 | NP_037531.2 | 6 | 1514 | |||
| GenBank mRNA | AF131218.1 | - | 5 | 1591 | ||
| AF195661.1 | - | 6 | 1509 | |||
| AK294257.1 | - | 6 | 1367 | |||
| AK308532.1 | - | 4 | 1154 | |||
| AL136698.1 | - | 6 | 1472 | |||
| AL833853.1 | - | 6 | 1546 | |||
| BC002882.2 | - | 6 | 1460 | 3941342 | ||
| BC007604.1 | - | 2 | 1048 | |||
| DQ167023.1 | - | 6 | 1483 | |||

CDIP1
Genomic Map