siRNA Details
| Gene Symbol: RHOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300886.1 | NP_001287815.1 | 2 | 302 | ||
| NM_014578.3 | NP_055393.1 | 4 | 500 | |||
| GenBank mRNA | AF498973.1 | - | 4 | 415 | ||
| BC001338.1 | - | 4 | 468 | 3461774 | ||
| BM761362.1 | - | 4 | 500 | |||
| CN305061.1 | - | 2 | 231 | |||

RHOD
Genomic Map