siRNA Details
| Gene Symbol: SLC2A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271711.1 | NP_001258640.1 | 9 | 1908 | ||
| NM_001271712.1 | NP_001258641.1 | 8 | 1684 | |||
| NM_014580.4 | NP_055395.2 | 10 | 2054 | |||
| NR_073416.1 | 7 | 1487 | ||||
| XM_006717084.3 | XP_006717147.1 | 10 | 1725 | |||
| XM_011518602.2 | XP_011516904.1 | 10 | 1970 | |||
| XM_011518603.2 | XP_011516905.1 | 8 | 1709 | |||
| XM_011518604.2 | XP_011516906.1 | 8 | 1633 | |||
| GenBank mRNA | AB209112.1 | - | 5278 | |||
| AF289587.1 | - | 9 | 1816 | |||
| BC019043.2 | - | 10 | 2006 | 4641145 | ||

SLC2A8
Genomic Map