siRNA Details
| Gene Symbol: GLTSCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015711.3 | NP_056526.3 | 6 | 1225 | ||
| XM_005258833.4 | XP_005258890.1 | 6 | 1268 | |||
| XM_006723180.3 | XP_006723243.1 | 6 | 1268 | |||
| XM_011526882.2 | XP_011525184.1 | 2 | 2235 | |||
| XM_011526883.2 | XP_011525185.1 | 6 | 1268 | |||
| GenBank mRNA | AF182077.1 | - | 1 | 1241 | ||
| BC032065.1 | - | 6 | 1225 | |||

GLTSCR1
Genomic Map